missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Tri-n-butyltin chloride, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1074618

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA1074618


Découvrez plus d'offres spéciales

Description

Description

Tri-n-butyltin chloride is used as an endocrine disruptor as well as an inhibitor for the V-ATPases. It is also used as a rodent-repellent for cable coatings. Further, it is used in hot end glass coating. It also employed as a corrosion remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyltin chloride is used as an endocrine disruptor as well as an inhibitor for the V-ATPases. It is also used as a rodent-repellent for cable coatings. Further, it is used in hot end glass coating. It also employed as a corrosion remover.

Solubility
Miscible with alcohol, heptane, benzene and toluene. Immiscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Spécifications

Spécifications

Tri-n-butyltin chloride
-19°C
171°C to 173°C (25 mmHg)
C12H27ClSn
MFCD00000521
UN2788
tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chloride
GCTFWCDSFPMHHS-UHFFFAOYSA-M
tributyl(chloro)stannane
15096
325.51
1461-22-9
1.2
120°C (248°F)
1.491
50 g
3535715
Miscible with alcohol,heptane,benzene and toluene. Immiscible with water.
CCCC[Sn](CCCC)(CCCC)Cl
325.508
CHEBI:79734
96%

Identifiants chimiques

1461-22-9
325.508
GCTFWCDSFPMHHS-UHFFFAOYSA-M
15096
tributyl(chloro)stannane
C12H27ClSn
MFCD00000521
tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chloride
CHEBI:79734
CCCC[Sn](CCCC)(CCCC)Cl
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H360FD-H372

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 215-958-7

missing translation for 'rtecsNumber' : WH6820000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Offres spéciales

Offres spéciales

RUO – Research Use Only