Thermo Scientific™ Tri-Sil BP (BSA:Pyridine) Reagent
Employ a reagent-solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil BP (BSA:Pyridine) Reagent,
Supplier:  Thermo Scientific™ TS49012
Catalog No. PI49012
Explore available promotions
Description
Description
Employ a reagent-solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil BP (BSA:Pyridine) Reagent, Composed of BSA and high-purity pyridine, this silylation reagent formulation derivatizes alcohols, phenols, organic acids, aromatic amides, and amines for GC analysis. It is excellent for use with unhindered steriods.
Reacts with a Wide Range of Compounds
Alcohols, phenols, some enols, and other hydroxyl/polyhydroxyl compounds to form trimethylsilyl ethers
Organic acids to form trimethylsilyl esters
Aromatic amides to form N-trimethylsilyl derivatives
Amines to form N-trimethylsilyl derivatives
Unhindered steroids
Not recommended for use with carbohydrates.
Specifications
Specifications
GC
Corrosive, Flammable
25 mL
65 - 80, 20 - 25%
2°C to 8°C
Pyridine, Trimethylsilyl N-trimethylsilylacetamidate
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.