Learn More
Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent
Employ a reagent-catalyst solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent,
$589.76 - $1064.44
Specifications
Standard Concentration | 65 - 80, 10 - 20, 5 - 7% |
---|---|
For Use With | GC |
Hazards | Corrosive, Flammable |
Storage | 2°C to 8°C |
Standard Components | Pyridine, Hexamethyldisilazane, Chlorotrimethylsilane |
Description
Employ a reagent-catalyst solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent, Composed of HMDS, TMCS and high-purity pyridine, it rapidly produces TMS derivatives of polar compounds for GC analysis and biochemical synthesis. This versatile reagent is ideally suited for GC determinations of a range of compounds including sugars, alcohols, phenols, steroids, sterols, organic acids, and some amines.
- Derivatizes carbohydrates, phenols, steroids, sterols, organic acids, alcohols and some amines
- Useful for rapid production of TMS derivatives of polar compounds for gas chromatographic determination and biochemical synthesis
- Versatility, speed and ease of use of Tri-Suil HTP Reagent has made it the most widely used silylation formulation
Specifications
65 - 80, 10 - 20, 5 - 7% | |
Corrosive, Flammable | |
Pyridine, Hexamethyldisilazane, Chlorotrimethylsilane |
GC | |
2°C to 8°C |
For Research Use Only. Not for use in diagnostic procedures.