missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent
Employ a catalyzed silylation reagent formulation, for derivatization of hydroxyls with the Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent.
Supplier: Thermo Scientific™ TS49016
Description
Employ a catalyzed silylation reagent formulation, for derivatization of hydroxyls with the Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent. Composed of TMSI/BSA/TMCS (3:3:2), this catalyzed silylation reagent formulation typically reacts with hydroxyls in a short period of time at 60–80 °C to form TMS ethers. Highly hindered hydroxyls may require several hours.
- Converts all classes of hydroxyl groups to TMS ethers
- Under usual conditions, the reaction is complete in a short period of time at 60 to 80°C, although very hindered hydroxyls may require several hours
Specifications
GC | |
Toxic | |
10 x 1 mL ampules |
25 - 45, 25 - 45, 20 - 25% | |
Room Temperature | |
Trimethylsilyl N-trimethylsilylacetamidate, 1-(Trimethylsilyl)-1 H-imidazole, Chlorotrimethylsilane |
For Research Use Only. Not for use in diagnostic procedures.