Learn More
Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent
Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent.
$1239.53 - $3184.79
Specifications
Standard Concentration | 65 - 80, 20 - 25% |
---|---|
For Use With | GC |
Storage | 2°C to 8°C |
Standard Components | Pyridine, 1-(Trimethylsilyl)-1 H-imidazole |
Description
Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent. Comprising TMSI in pyridine, this reagent silylates alcohols and phenols, organic acids, hydroxylamines, amino acids, carbohydrates, flavonoids, glycols and polyglycols, nucleotides, steroids, hydroxyl acids, barbituates, narcotics, indoles, and vitamins.
Additonal Product Information
Does not react with amines
May be used in the presence of water as long as there is enough reagent present to react with both the water and the sample.
Specifications
65 - 80, 20 - 25% | |
GC | |
2°C to 8°C | |
Pyridine, 1-(Trimethylsilyl)-1 H-imidazole |
For Research Use Only. Not for use in diagnostic procedures.