Spectrum™ Trial Size Kits for Biotech-Grade CE Dialysis Membrane Tubing (1 meter), MWCO: 1000 kd
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
Supplier: Spectrum™ 131492T
Includes:
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Description
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
Kits are great for comparing membrane MWCO performance to optimize separation or purification. Provides a 1m length of Biotech-Grade CE membrane tubing, a pair of universal tubing closures and 5 picks for opening membrane tubing. Tubing is wet packaged in 0.05% sodium azide preservative that easily rinses out with water.
- Rigidly controlled porosity for better selectivity and separations than Standard Grade RC
- Ultrapure, no heavy metal or sulfiide impurities
- Low protein binding
- No glycerin removal, cleaning or pre-treatment required
Specifications
1000 kDa | |
20 mm | |
3.1 mL/cm vol./length | |
1 m Tubing/Kit |