missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triallyl Cyanurate 98.0+%, TCI America™
$57.27 - $256.75
Chemical Identifiers
CAS | 101-37-1 |
---|---|
Molecular Formula | C12H15N3O3 |
Molecular Weight (g/mol) | 249.27 |
MDL Number | MFCD00006049 |
InChI Key | BJELTSYBAHKXRW-UHFFFAOYSA-N |
Synonym | triallyl cyanurate, 2,4,6-triallyloxy-1,3,5-triazine, triallyl cyanaurate, activator oc, rhenofit tac, cyanuric acid triallyl ester, perkalink 300, tripropargyl cyanurate, perkalink 300-50d, 1,3,5-triazine, 2,4,6-tris 2-propenyloxy |
PubChem CID | 7555 |
IUPAC Name | 2,4,6-tris(prop-2-enoxy)-1,3,5-triazine |
SMILES | C=CCOC1=NC(=NC(=N1)OCC=C)OCC=C |
Chemical Identifiers
101-37-1 | |
249.27 | |
BJELTSYBAHKXRW-UHFFFAOYSA-N | |
7555 | |
C=CCOC1=NC(=NC(=N1)OCC=C)OCC=C |
C12H15N3O3 | |
MFCD00006049 | |
triallyl cyanurate, 2,4,6-triallyloxy-1,3,5-triazine, triallyl cyanaurate, activator oc, rhenofit tac, cyanuric acid triallyl ester, perkalink 300, tripropargyl cyanurate, perkalink 300-50d, 1,3,5-triazine, 2,4,6-tris 2-propenyloxy | |
2,4,6-tris(prop-2-enoxy)-1,3,5-triazine |
Specifications
101-37-1 | |
162°C | |
MFCD00006049 | |
3077 | |
BJELTSYBAHKXRW-UHFFFAOYSA-N | |
2,4,6-tris(prop-2-enoxy)-1,3,5-triazine | |
7555 | |
≥98.0% (GC) | |
Triallyl Cyanurate |
White | |
C12H15N3O3 | |
25 g | |
triallyl cyanurate, 2,4,6-triallyloxy-1,3,5-triazine, triallyl cyanaurate, activator oc, rhenofit tac, cyanuric acid triallyl ester, perkalink 300, tripropargyl cyanurate, perkalink 300-50d, 1,3,5-triazine, 2,4,6-tris 2-propenyloxy | |
C=CCOC1=NC(=NC(=N1)OCC=C)OCC=C | |
249.27 | |
249.27 | |
Crystalline Lumps |
Safety and Handling
EINECSNumber : (5)-1059
RTECSNumber : XZ2080000
TSCA : Yes