missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triamcinolone acetonide, 98+%, Thermo Scientific Chemicals

$92.96 - $238.27

Chemical Identifiers

CAS 76-25-5
Molecular Formula C24H31FO6
Molecular Weight (g/mol) 434.504
MDL Number MFCD00056834
InChI Key YNDXUCZADRHECN-JNQJZLCISA-N
Synonym triamcinolone acetonide, azmacort, nasacort aq, aristoderm, aristogel, kenalone, solodelf, tramacin, triaceton, tricinolon
PubChem CID 6436
ChEBI CHEBI:71418
SMILES CC1(OC2CC3C4CCC5=CC(=O)C=CC5(C4(C(CC3(C2(O1)C(=O)CO)C)O)F)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63548MD
View Documents
Thermo Scientific Chemicals
J63548MD
250 mg
Each for $92.96
 
AAJ6354803
View Documents
Thermo Scientific Chemicals
J6354803
1 g
Each for $238.27
 
Description

Description

Triamcinolone acetonide suppresses allergic, inflammatory, and immune responses. Intravitreal administration of triamcinolone acetonide is used against diabetic macular edema. It is also used to relieve the discomfort of mouth sores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in DMSO or ethanol. Slightly soluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, acids, heat.
Specifications

Chemical Identifiers

76-25-5
434.504
YNDXUCZADRHECN-JNQJZLCISA-N
6436
CC1(OC2CC3C4CCC5=CC(=O)C=CC5(C4(C(CC3(C2(O1)C(=O)CO)C)O)F)C)C
C24H31FO6
MFCD00056834
triamcinolone acetonide, azmacort, nasacort aq, aristoderm, aristogel, kenalone, solodelf, tramacin, triaceton, tricinolon
CHEBI:71418

Specifications

76-25-5
White
MFCD00056834
14,9596
Soluble in DMSO or ethanol; Slightly soluble in water.
CC1(OC2CC3C4CCC5=CC(=O)C=CC5(C4(C(CC3(C2(O1)C(=O)CO)C)O)F)C)C
6436
434.5
Powder
274°C to 278°C
C24H31FO6
250 mg
triamcinolone acetonide, azmacort, nasacort aq, aristoderm, aristogel, kenalone, solodelf, tramacin, triaceton, tricinolon
YNDXUCZADRHECN-JNQJZLCISA-N
434.504
CHEBI:71418
≥98%
Triamcinolone acetonide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H302
May damage fertility or the unborn child.
Harmful if swallowed.

P201-P202-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H360

EINECSNumber : 200-948-7

RTECSNumber : TU3920000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only