Eisco™ Triangular Base w/ Rod, Base 6.7 in. wide, Rod 19.7 in. long, .4 in. (11mm) dia
6.75 in. Cast Iron black or blue base paired with a Steel rod 20 in. 11mm in diameter. Rod is threaded 10 x 1.5mm Base provides 3 points of support for stability.
Supplier:  Eisco™ TRISTD1
Catalog No. S23770
Explore available promotions
Description
Description
  • Rod and base set consisting of a Triangular Tripod Retort Base made of corrosion-resistant, powder-coated pressed steel and 50 cm rod made of steel
  • The base weighs 1.8 lb., providing superb stability for experiments and setups - The included rod measures 20 in. tall and is 12 mm in diameter
  • Base features a tapped hole, which fits the included rod, and can accommodate other rods with a 10 mm x 1.5 mm metric thread
  • The durable and sturdy construction of this set makes it an excellent choice for science classrooms and basic professional laboratory setups
Specifications
Specifications
Cast Iron (Base), Steel (Rod)
19.7 in.
0.4 in.
Stands and Rings
25.4 mm
6.7 in.
11 mm
Triangular Base
Product Suggestions
Product Suggestions