Learn More
United Scientific™ Triangular File with Handle
This Triangular File with Handle is used to score glass tubing.
Supplier:  United Scientific™ TRF004
 View more versions of this product
Catalog No. S66970
Only null left
Explore available promotions
Alert:
Prop 65
Description
Description
  • Use to score glass before cutting
  • Files have tapered shape and are mounted on a hardwood handle for easy gripping
  • Available in four sizes
Specifications
Specifications
Triangular File
Steel
No
Hardwood
17.8 cm
Steel
Scoring Glass Tubing for Cutting
Round
Triangular
Used with glass to score before cutting
7 in.
10.2 cm
4 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Warning: Cancer and Reproductive Harm