missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributyl(cyanomethyl)phosphonium Chloride 98.0+%, TCI America™
$96.94 - $197.59
Chemical Identifiers
| CAS | 82358-61-0 |
|---|---|
| Molecular Formula | C14H29ClNP |
| Molecular Weight (g/mol) | 277.817 |
| MDL Number | MFCD00671508 |
| InChI Key | JCHWNYYARSRCKZ-UHFFFAOYSA-M |
| Synonym | (Cyanomethyl)tributylphosphonium Chloride |
| PubChem CID | 11594300 |
| IUPAC Name | tributyl(cyanomethyl)phosphanium;chloride |
| SMILES | CCCC[P+](CCCC)(CCCC)CC#N.[Cl-] |
Chemical Identifiers
| 82358-61-0 | |
| 277.817 | |
| JCHWNYYARSRCKZ-UHFFFAOYSA-M | |
| 11594300 | |
| CCCC[P+](CCCC)(CCCC)CC#N.[Cl-] |
| C14H29ClNP | |
| MFCD00671508 | |
| (Cyanomethyl)tributylphosphonium Chloride | |
| tributyl(cyanomethyl)phosphanium;chloride |
Specifications
| 82358-61-0 | |
| White | |
| MFCD00671508 | |
| 3439 | |
| JCHWNYYARSRCKZ-UHFFFAOYSA-M | |
| tributyl(cyanomethyl)phosphanium;chloride | |
| 11594300 | |
| ≥98.0% (T) | |
| Tributyl(cyanomethyl)phosphonium Chloride |
| 97°C | |
| C14H29ClNP | |
| 10 g | |
| (Cyanomethyl)tributylphosphonium Chloride | |
| CCCC[P+](CCCC)(CCCC)CC#N.[Cl-] | |
| 277.817 | |
| 277.82 | |
| Crystalline Powder |
Safety and Handling
TSCA : No