missing translation for 'onlineSavingsMsg'
Learn More

Tributylhexadecylphosphonium Bromide 98.0+%, TCI America™

$272.16 - $272.16

Chemical Identifiers

CAS 14937-45-2
Molecular Formula C28H60BrP
Molecular Weight (g/mol) 507.67
MDL Number MFCD00011775
InChI Key RYVBINGWVJJDPU-UHFFFAOYSA-M
Synonym tributylhexadecylphosphonium bromide, tributyl hexadecyl phosphonium bromide, cetyltributylphosphonium bromide, tributyl hexadecyl phosphanium bromide, phosphonium, tributylhexadecyl-, bromide, hexadecyltributylphosphonium bromide, phosphonium, tributylhexadecyl-, bromide 1:1, hexadecyltri-n-butylphosphonium bromide, n-hexadecyl tri-n-butylphosphonium bromide, tbhdpb
PubChem CID 84716
IUPAC Name tributyl(hexadecyl)phosphanium bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H104725G
View Documents
TCI America
H104725G
25 g
Each for $272.16
Only null left
 
Description

Description

Specifications

Chemical Identifiers

14937-45-2
507.67
RYVBINGWVJJDPU-UHFFFAOYSA-M
84716
[Br-].CCCCCCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC
C28H60BrP
MFCD00011775
tributylhexadecylphosphonium bromide, tributyl hexadecyl phosphonium bromide, cetyltributylphosphonium bromide, tributyl hexadecyl phosphanium bromide, phosphonium, tributylhexadecyl-, bromide, hexadecyltributylphosphonium bromide, phosphonium, tributylhexadecyl-, bromide 1:1, hexadecyltri-n-butylphosphonium bromide, n-hexadecyl tri-n-butylphosphonium bromide, tbhdpb
tributyl(hexadecyl)phosphanium bromide

Specifications

14937-45-2
White
MFCD00011775
tributylhexadecylphosphonium bromide, tributyl hexadecyl phosphonium bromide, cetyltributylphosphonium bromide, tributyl hexadecyl phosphanium bromide, phosphonium, tributylhexadecyl-, bromide, hexadecyltributylphosphonium bromide, phosphonium, tributylhexadecyl-, bromide 1:1, hexadecyltri-n-butylphosphonium bromide, n-hexadecyl tri-n-butylphosphonium bromide, tbhdpb
[Br-].CCCCCCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC
507.67
507.67
Crystals
61°C
C28H60BrP
25 g
RYVBINGWVJJDPU-UHFFFAOYSA-M
tributyl(hexadecyl)phosphanium bromide
84716
≥98.0% (T)
Tributylhexadecylphosphonium Bromide
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'