missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Tributyrin | |
-75°C | |
190°C | |
MFCD00009392 | |
tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC | |
302.367 | |
CHEBI:35020 | |
≥98.0% (GC) |
60-01-5 | |
Yellow | |
C15H26O6 | |
500 mL | |
UYXTWWCETRIEDR-UHFFFAOYSA-N | |
2,3-di(butanoyloxy)propyl butanoate | |
6050 | |
302.37 | |
Liquid |
Chemical Identifiers
60-01-5 | |
302.367 | |
UYXTWWCETRIEDR-UHFFFAOYSA-N | |
6050 | |
2,3-di(butanoyloxy)propyl butanoate |
C15H26O6 | |
MFCD00009392 | |
tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin | |
CHEBI:35020 | |
CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC |
Safety and Handling
EINECSNumber : (2)-0654&(2)-0666
RTECSNumber : ET7350000
TSCA : Yes