missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trichloroacetic anhydride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0084918

 View more versions of this product

Catalog No. AAL0084918


Explore available promotions

Description

Description

Trichloroacetic anhydride is used as a derivatization reagent involved in the analysis of amphetamine,methamphetamine and 3,4-methylenedioxymethamphetamine. It is also used in the determination of the metabolites of l-alpha-acetylmethadol (LAAM), such as noracetylmethadol, dinoracetylmethadol, methadol and normethadol by electron capture gas-liquid chromatography. It is used in the synthesis of trichloroacetonylpyridine by reacting with 2-methyl-pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroacetic anhydride is used as a derivatization reagent involved in the analysis of amphetamine,methamphetamine and 3,4-methylenedioxymethamphetamine. It is also used in the determination of the metabolites of l-alpha-acetylmethadol (LAAM), such as noracetylmethadol, dinoracetylmethadol, methadol and normethadol by electron capture gas-liquid chromatography. It is used in the synthesis of trichloroacetonylpyridine by reacting with 2-methyl-pyridine.

Solubility
Miscible with ether and acetic acid.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases with strong oxidizing agents.
Specifications

Specifications

Trichloroacetic anhydride
1.69
C4Cl6O3
MFCD00000793
UN3265
Moisture sensitive
Miscible with ether and acetic acid.
C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl
308.741
308.76
4124-31-6
98°C to 100°C (11 mmHg)
1.484
50 g
980355
trichloroacetic anhydride, acetic acid, trichloro-, anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acmc-1aliy, perchloroacetic anhydride, bis trichloroacetic anhydride, trichloroacetic acid anhydride, 2,2,2-trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate
MEFKFJOEVLUFAY-UHFFFAOYSA-N
(2,2,2-trichloroacetyl) 2,2,2-trichloroacetate
20079
95%

Chemical Identifiers

4124-31-6
308.741
MEFKFJOEVLUFAY-UHFFFAOYSA-N
20079
C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl
C4Cl6O3
MFCD00000793
trichloroacetic anhydride, acetic acid, trichloro-, anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acmc-1aliy, perchloroacetic anhydride, bis trichloroacetic anhydride, trichloroacetic acid anhydride, 2,2,2-trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate
(2,2,2-trichloroacetyl) 2,2,2-trichloroacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: I

EINECSNumber : 223-925-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only