missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trichloroethylene, ACS, 99.5% min, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 019401K7

 View more versions of this product

Catalog No. AA19401K7


Explore available promotions

Description

Description

Trichloroethylene is used as solvent for paints, dyes, waxes, oils, fats and resins, as an additive in polymerization reactions, as a reactant to prepare various halohydrocarbons (1,1,1,2-tetrafluoroethane) and other chemicals, as a coolant for cutting tantalum and acts as good heat transfer medium. It is used in fire extinguisher fluids and in the recovery of oils and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Trichloroethylene
79-01-6
1.46 g/mL
None
Chloroform-like
1.476
4 L
1736782
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
XSTXAVWGXDQKEL-UHFFFAOYSA-N
1,1,2-trichloroethene
6575
131.39
ACS
For ACS analysis
-85°C
87°C
9.5% min.
C2HCl3
MFCD00000838
UN1710
14,9639
Miscible with ether,alcohol,chloroformMiscible with ether,ethanol and chloroform; Very slightly miscible with water.
C(=C(Cl)Cl)Cl
131.38
CHEBI:16602
≤99.5%
Liquid

Chemical Identifiers

79-01-6
131.38
XSTXAVWGXDQKEL-UHFFFAOYSA-N
6575
1,1,2-trichloroethene
C2HCl3
MFCD00000838
trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
CHEBI:16602
C(=C(Cl)Cl)Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H341-H315-H319-H336
May cause cancer.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.

P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c

H315-H317-H319-H336-H341-H350-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TRICHLOROETHYLENE

EINECSNumber : 201-167-4

RTECSNumber : KX4550000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only