missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trichostatin A 98.0+%, TCI America™

Supplier:  TCI America T247750MG

 View more versions of this product

Catalog No. T247750MG


Explore available promotions

Description

Description

Specifications

Specifications

Trichostatin A
144°C
C17H22N2O3
50 mg
RTKIYFITIVXBLE-WKWSCTOINA-N
(2E,4E)-7-[4-(dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide
444732
302.37
Crystalline Powder
58880-19-6
White-Yellow
MFCD03848392
trichostatin a, trichostatin a tsa, trichostatin, tsa, unii-3x2s926l3z, 2e,4e,6r-7-4-dimethylamino phenyl-n-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide, gnf-pf-1011, 2,4-heptadienamide, 7-4-dimethylamino phenyl-n-hydroxy-4,6-dimethyl-7-oxo, 7-4-dimethylamino phenyl-n-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide, r-e,e-7-4-dimethylamino phenyl-n-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide
CC(\C=C(/C)\C=C\C(=O)NO)C(=O)C1=CC=C(C=C1)N(C)C
302.37
CHEBI:46024
≥98.0% (HPLC)

Chemical Identifiers

58880-19-6
302.37
RTKIYFITIVXBLE-WKWSCTOINA-N
444732
(2E,4E)-7-[4-(dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide
C17H22N2O3
MFCD03848392
trichostatin a, trichostatin a tsa, trichostatin, tsa, unii-3x2s926l3z, 2e,4e,6r-7-4-dimethylamino phenyl-n-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide, gnf-pf-1011, 2,4-heptadienamide, 7-4-dimethylamino phenyl-n-hydroxy-4,6-dimethyl-7-oxo, 7-4-dimethylamino phenyl-n-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide, r-e,e-7-4-dimethylamino phenyl-n-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide
CHEBI:46024
CC(\C=C(/C)\C=C\C(=O)NO)C(=O)C1=CC=C(C=C1)N(C)C
Safety and Handling

Safety and Handling

RTECSNumber : MI5215000

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions