missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tricine Buffer, MP Biomedicals
Supplier: MP Biomedicals Inc 0210311280
Specifications
Tricine Buffer | |
5704-04-1 | |
Powder | |
C6H13NO5 | |
N-Tris-[hydroxymethyl]methylglycine, N-[2-Hydroxy-1, 1-bis(hydroxymethyl) ethyl] glycine | |
C(C(=O)O)NC(CO)(CO)CO | |
179.17 | |
CHEBI:39063 |
Cell Biology and Analysis, Molecular Biology | |
White | |
100 g | |
MFCD00004277 | |
SEQKRHFRPICQDD-UHFFFAOYSA-N | |
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid | |
79784 | |
179.2 |
Chemical Identifiers
5704-04-1 | |
179.17 | |
SEQKRHFRPICQDD-UHFFFAOYSA-N | |
79784 | |
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid |
C6H13NO5 | |
MFCD00004277 | |
N-Tris-[hydroxymethyl]methylglycine, N-[2-Hydroxy-1, 1-bis(hydroxymethyl) ethyl] glycine | |
CHEBI:39063 | |
C(C(=O)O)NC(CO)(CO)CO |
Safety and Handling
EINECSNumber : 227-193-6
Recommended Storage : Store at room temperature (15° to 30°C).