missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tricine Buffer, MP Biomedicals

Supplier:  MP Biomedicals Inc 0210311283

 View more versions of this product

Catalog No. ICN10311283


Explore available promotions

Description

Description

Specifications

Specifications

Tricine Buffer
5704-04-1
Powder
C6H13NO5
N-Tris-[hydroxymethyl]methylglycine, N-[2-Hydroxy-1, 1-bis(hydroxymethyl) ethyl] glycine
C(C(=O)O)NC(CO)(CO)CO
179.17
CHEBI:39063
Cell Biology and Analysis, Molecular Biology
White
250 g
MFCD00004277
SEQKRHFRPICQDD-UHFFFAOYSA-N
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
79784
179.2

Chemical Identifiers

5704-04-1
179.17
SEQKRHFRPICQDD-UHFFFAOYSA-N
79784
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C6H13NO5
MFCD00004277
N-Tris-[hydroxymethyl]methylglycine, N-[2-Hydroxy-1, 1-bis(hydroxymethyl) ethyl] glycine
CHEBI:39063
C(C(=O)O)NC(CO)(CO)CO
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-193-6

Recommended Storage : Store at room temperature (15° to 30°C).

SDS
Documents

Documents

Promotions

Promotions