missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tricyclo[5.2.1.0(2,6)]decan-8-one 95.0+%, TCI America™

Supplier:  TCI America T1321500ML

 View more versions of this product

Catalog No. T1321500ML


Explore available promotions

Description

Description

Specifications

Specifications

Tricyclo[5.2.1.0(2,6)]decan-8-one
Colorless
132°C
MFCD00168034
OMIDXVJKZCPKEI-UHFFFAOYSA-N
150.221
150.22
Liquid
13380-94-4
7 (3g/L H2O soln. 20°C)
C10H14O
500 mL
C1CC2C(C1)C3CC2CC3=O
96509
≥95.0% (GC)

Chemical Identifiers

13380-94-4
150.221
OMIDXVJKZCPKEI-UHFFFAOYSA-N
C1CC2C(C1)C3CC2CC3=O
C10H14O
MFCD00168034
96509
Videos
Safety and Handling

Safety and Handling

RTECSNumber : PC0350000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions