missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tridecafluoroheptanoic Acid High Grade 98.0+%, TCI America™

$175.78 - $521.40

Chemical Identifiers

CAS 375-85-9
Molecular Formula C7HF13O2
Molecular Weight (g/mol) 364.062
MDL Number MFCD00039604
InChI Key ZWBAMYVPMDSJGQ-UHFFFAOYSA-N
Synonym perfluoroheptanoic acid, tridecafluoroheptanoic acid, perfluoroenanthic acid, perfluoro-n-heptanoic acid, heptanoic acid, tridecafluoro, tridecafluoro-1-heptanoic acid, heptanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoro, n-perfluoroheptanoic acid, tridecafluoroenanthic acid, acmc-1agq8
PubChem CID 67818
ChEBI CHEBI:35547
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoic acid
SMILES C(=O)(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A57211G
View Documents
TCI America
A57211G
1 g
Each for $175.78
 
A57215G
View Documents
TCI America
A57215G
5 g
Each for $521.40
 
Description

Description

Specifications

Chemical Identifiers

375-85-9
364.062
ZWBAMYVPMDSJGQ-UHFFFAOYSA-N
67818
2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoic acid
C7HF13O2
MFCD00039604
perfluoroheptanoic acid, tridecafluoroheptanoic acid, perfluoroenanthic acid, perfluoro-n-heptanoic acid, heptanoic acid, tridecafluoro, tridecafluoro-1-heptanoic acid, heptanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoro, n-perfluoroheptanoic acid, tridecafluoroenanthic acid, acmc-1agq8
CHEBI:35547
C(=O)(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

375-85-9
177°C
MFCD00039604
3261
ZWBAMYVPMDSJGQ-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoic acid
67818
364.06
Crystalline Powder
White
C7HF13O2
1 g
perfluoroheptanoic acid, tridecafluoroheptanoic acid, perfluoroenanthic acid, perfluoro-n-heptanoic acid, heptanoic acid, tridecafluoro, tridecafluoro-1-heptanoic acid, heptanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoro, n-perfluoroheptanoic acid, tridecafluoroenanthic acid, acmc-1agq8
C(=O)(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O
364.062
CHEBI:35547
≥98.0% (T)
Tridecafluoroheptanoic Acid High Grade [Ion-Pair Reagent for LC-MS]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1182&(2)-2659

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'