missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tridecafluorohexyl Iodide 98.0+%, TCI America™
$44.73 - $147.25
Chemical Identifiers
| CAS | 355-43-1 |
|---|---|
| Molecular Formula | C6F13I |
| Molecular Weight (g/mol) | 445.95 |
| MDL Number | MFCD00001063 |
| InChI Key | BULLJMKUVKYZDJ-UHFFFAOYSA-N |
| Synonym | perfluorohexyl iodide, perfluoro-1-iodohexane, 1-iodoperfluorohexane, perfluoro-n-hexyl iodide, tridecafluorohexyl iodide, unii-a2a17jug4r, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodo, tridecafluoro-1-iodohexane, a2a17jug4r, hexane, tridecafluoro-6-iodo |
| PubChem CID | 67733 |
| IUPAC Name | 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane |
| SMILES | C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F |
Chemical Identifiers
| 355-43-1 | |
| 445.95 | |
| BULLJMKUVKYZDJ-UHFFFAOYSA-N | |
| 67733 | |
| C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F |
| C6F13I | |
| MFCD00001063 | |
| perfluorohexyl iodide, perfluoro-1-iodohexane, 1-iodoperfluorohexane, perfluoro-n-hexyl iodide, tridecafluorohexyl iodide, unii-a2a17jug4r, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodo, tridecafluoro-1-iodohexane, a2a17jug4r, hexane, tridecafluoro-6-iodo | |
| 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane |
Specifications
| 355-43-1 | |
| Yellow | |
| C6F13I | |
| 5 g | |
| BULLJMKUVKYZDJ-UHFFFAOYSA-N | |
| 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane | |
| 67733 | |
| ≥98.0% (GC) | |
| Tridecafluorohexyl Iodide |
| -45°C | |
| 117°C | |
| MFCD00001063 | |
| perfluorohexyl iodide, perfluoro-1-iodohexane, 1-iodoperfluorohexane, perfluoro-n-hexyl iodide, tridecafluorohexyl iodide, unii-a2a17jug4r, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodo, tridecafluoro-1-iodohexane, a2a17jug4r, hexane, tridecafluoro-6-iodo | |
| C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F | |
| 445.95 | |
| 445.95 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0090
TSCA : Yes