missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tridecylbenzene 99.0+%, TCI America™
$93.43 - $261.24
Chemical Identifiers
CAS | 123-02-4 |
---|---|
Molecular Formula | C19H32 |
Molecular Weight (g/mol) | 260.465 |
MDL Number | MFCD00008980 |
InChI Key | MCVUKOYZUCWLQQ-UHFFFAOYSA-N |
Synonym | 1-phenyltridecane, benzene, tridecyl, n-tridecylbenzene, tridane, tridecane, 1-phenyl, benzene, c10-16-alkyl derivs., detergent alkylate no. 5, unii-e6k14656yo, c8-c16 alkylbenzene, benzene, mono-c12-14-alkyl derivs. |
PubChem CID | 31238 |
IUPAC Name | tridecylbenzene |
SMILES | CCCCCCCCCCCCCC1=CC=CC=C1 |
Chemical Identifiers
123-02-4 | |
260.465 | |
MCVUKOYZUCWLQQ-UHFFFAOYSA-N | |
31238 | |
CCCCCCCCCCCCCC1=CC=CC=C1 |
C19H32 | |
MFCD00008980 | |
1-phenyltridecane, benzene, tridecyl, n-tridecylbenzene, tridane, tridecane, 1-phenyl, benzene, c10-16-alkyl derivs., detergent alkylate no. 5, unii-e6k14656yo, c8-c16 alkylbenzene, benzene, mono-c12-14-alkyl derivs. | |
tridecylbenzene |
Specifications
123-02-4 | |
Colorless | |
C19H32 | |
5 mL | |
MCVUKOYZUCWLQQ-UHFFFAOYSA-N | |
tridecylbenzene | |
31238 | |
≥99.0% (GC) | |
Tridecylbenzene |
10°C | |
346°C | |
MFCD00008980 | |
1-phenyltridecane, benzene, tridecyl, n-tridecylbenzene, tridane, tridecane, 1-phenyl, benzene, c10-16-alkyl derivs., detergent alkylate no. 5, unii-e6k14656yo, c8-c16 alkylbenzene, benzene, mono-c12-14-alkyl derivs. | |
CCCCCCCCCCCCCC1=CC=CC=C1 | |
260.465 | |
260.47 | |
Liquid |
Safety and Handling
EINECSNumber : (3)-0021
TSCA : Yes