missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Triethanolamine, 97 to 98%, MP Biomedicals™

Triethanolamine ≥97%, Liquid

$93.18 - $136.18

Identifiants chimiques

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
ICN15216380
Afficher les documents
MP Biomedicals Inc
0215216380
100 mL
chaque for $93.18
 
MP215216390
Afficher les documents
MP Biomedicals Inc
0215216390
500 mL
chaque for $136.18
 
Description

Description

  • It is used as an emulsifier and a surfactant. It finds application in common products like dishwashing liquids, general cleaners, hand cleaners, shaving cream, metalworking fluids, liquid laundry detergents, paints and printing inks.
  • It is used for the neutralization of fatty acids and adjusting buffer pH as well as utilized to dissolve oils. It acts as an effective complexing agent in electroless plating.
  • It is used in hair care products like hair dyes and wave sets.
  • Protein purification reagent Triethanolamine acts as an organic additive in the grinding of cement clinker and as a complexing agent for aluminum ions in aqueous solutions.
Spécifications

Identifiants chimiques

9005-65-6
604.82
HDTIFOGXOGLRCB-KTKRTIGZNA-N
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
MFCD00082107
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate

Spécifications

9005-65-6
1.124g/cm3 at 20°C
(C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
100 mL
HDTIFOGXOGLRCB-KTKRTIGZNA-N
2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
149.2
Triethanolamine
Colorless to Yellow
97 to 98%
MFCD00082107
Daltogen, Nitrilo-2, 2, 2-Triethanol
CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
604.82
Liquid
Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'einecsNumber' : 203-049-8

missing translation for 'storageNote' : Store Desiccated.. Store Under Nitrogen. Protect from light

Recommended Storage : Store at Room Temperature (15° to 30°C)

FDS
missing translation for 'documents'

missing translation for 'documents'