missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethanolamine (Certified), Fisher Chemical™

Supplier:  Thermo Fisher Scientific T4071

 View more versions of this product

Catalog No. T407-1



Explore available promotions

Description

Description

Specifications

Specifications

Triethanolamine
>325°C
Yellow
10.5
≥99 %
C6H15NO3
MFCD00002855
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
C(CO)N(CCO)CCO
149.19
CHEBI:28621
<0.01mmHg at 20°C
Certified
102-71-6
21°C
1.125g/cm³
360°C
Glass Bottle
N(CH2CH2OH)3
1 L
GSEJCLTVZPLZKY-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]ethanol
7618
149.14
≥99%
Viscous Liquid

Chemical Identifiers

102-71-6
149.19
GSEJCLTVZPLZKY-UHFFFAOYSA-N
7618
2-[bis(2-hydroxyethyl)amino]ethanol
C6H15NO3
MFCD00002855
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
CHEBI:28621
C(CO)N(CCO)CCO
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes. May cause an allergic skin reaction. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:0

SDS Documents

Documents

Promotions

Promotions