missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethyl 2-phosphonopentanoate, 98%, Thermo Scientific Chemicals
$228.27 - $836.38
Chemical Identifiers
CAS | 35051-49-1 |
---|---|
Molecular Formula | C11H23O5P |
Molecular Weight (g/mol) | 266.274 |
MDL Number | MFCD00015163 |
InChI Key | BUPVIVDUPRDDFI-UHFFFAOYSA-N |
Synonym | triethyl 2-phosphonopentanoate, ethyl 2-diethoxyphosphoryl pentanoate, pentanoic acid, 2-diethoxyphosphinyl-, ethyl ester, valeric acid, 2-diethylphosphono-, ethyl ester, alpha-carbethoxybutylphosphonic acid, diethyl ester, triethyl 2-phosphonovalerate, phosphonic acid, alpha-carbethoxybutyl-, diethyl ester, ethyl 2-diethoxyphosphoryl pentanoate #, diethyl 1-ethoxycarbonyl butylphosphonate |
PubChem CID | 98244 |
IUPAC Name | ethyl 2-diethoxyphosphorylpentanoate |
SMILES | CCCC(C(=O)OCC)P(=O)(OCC)OCC |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
35051-49-1 | |
266.274 | |
BUPVIVDUPRDDFI-UHFFFAOYSA-N | |
98244 | |
CCCC(C(=O)OCC)P(=O)(OCC)OCC |
C11H23O5P | |
MFCD00015163 | |
triethyl 2-phosphonopentanoate, ethyl 2-diethoxyphosphoryl pentanoate, pentanoic acid, 2-diethoxyphosphinyl-, ethyl ester, valeric acid, 2-diethylphosphono-, ethyl ester, alpha-carbethoxybutylphosphonic acid, diethyl ester, triethyl 2-phosphonovalerate, phosphonic acid, alpha-carbethoxybutyl-, diethyl ester, ethyl 2-diethoxyphosphoryl pentanoate #, diethyl 1-ethoxycarbonyl butylphosphonate | |
ethyl 2-diethoxyphosphorylpentanoate |
Specifications
35051-49-1 | |
98°C (2 mmHg) | |
(C2H5O)2P(O)[CH(C3H7)CO2C2H5] | |
2 g | |
BUPVIVDUPRDDFI-UHFFFAOYSA-N | |
ethyl 2-diethoxyphosphorylpentanoate | |
98244 | |
98% | |
Triethyl 2-phosphonopentanoate |
Colorless | |
C11H23O5P | |
MFCD00015163 | |
triethyl 2-phosphonopentanoate, ethyl 2-diethoxyphosphoryl pentanoate, pentanoic acid, 2-diethoxyphosphinyl-, ethyl ester, valeric acid, 2-diethylphosphono-, ethyl ester, alpha-carbethoxybutylphosphonic acid, diethyl ester, triethyl 2-phosphonovalerate, phosphonic acid, alpha-carbethoxybutyl-, diethyl ester, ethyl 2-diethoxyphosphoryl pentanoate #, diethyl 1-ethoxycarbonyl butylphosphonate | |
CCCC(C(=O)OCC)P(=O)(OCC)OCC | |
266.274 | |
266.27 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only