missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethyl methanetricarboxylate, 98%, Thermo Scientific Chemicals

$53.88 - $150.84

Chemical Identifiers

CAS 6279-86-3
Molecular Formula C10H16O6
Molecular Weight (g/mol) 232.232
MDL Number MFCD00009150
InChI Key AGZPNUZBDCYTBB-UHFFFAOYSA-N
Synonym tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane
PubChem CID 80471
IUPAC Name triethyl methanetricarboxylate
SMILES CCOC(=O)C(C(=O)OCC)C(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0835606
View Documents
Thermo Scientific Chemicals
L0835606
5 g
Each for $53.88
 
AAL0835614
View Documents
Thermo Scientific Chemicals
L0835614
25 g
Each for $150.84
 
Description

Description

Methanetricarboxylates have been used as blocked malonic esters, restricting reaction with alkyl halides to monoalkylation. The unwanted carboxyl group can be removed by treatment with Na alkoxide, LDA or BCl3. Provides a useful 2-carbon chain extension method that adds to Michael acceptors under phase-transfer conditions. Triethyl Methanetricarboxylate is used in the synthesis of novel inhibitors of Hsp90. It is also used in the preparation of novel dihydroquinoline-3-carboxylic acids that function as HIV-1 integrase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methanetricarboxylates have been used as “blocked” malonic esters, restricting reaction with alkyl halides to monoalkylation. The unwanted carboxyl group can be removed by treatment with Na alkoxide, LDA or BCl3. Provides a useful 2-carbon chain extension method that adds to Michael acceptors under phase-transfer conditions. Triethyl Methanetricarboxylate is used in the synthesis of novel inhibitors of Hsp90. It is also used in the preparation of novel dihydroquinoline-3-carboxylic acids that function as HIV-1 integrase inhibitors.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

6279-86-3
232.232
AGZPNUZBDCYTBB-UHFFFAOYSA-N
80471
CCOC(=O)C(C(=O)OCC)C(=O)OCC
C10H16O6
MFCD00009150
tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane
triethyl methanetricarboxylate

Specifications

6279-86-3
1.095
>110°C (230°F)
1.4255
5 g
tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane
AGZPNUZBDCYTBB-UHFFFAOYSA-N
triethyl methanetricarboxylate
80471
98%
26°C to 29°C
253°C
C10H16O6
MFCD00009150
1793645
Insoluble in water.
CCOC(=O)C(C(=O)OCC)C(=O)OCC
232.232
232.23
Triethyl methanetricarboxylate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 228-477-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only