missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthoacetate, 97%

CAS: 78-39-7 | C8H18O3 | 162.229 g/mol

$83.14 - $727.30

Chemical Identifiers

CAS 78-39-7
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD00009223
InChI Key NDQXKKFRNOPRDW-UHFFFAOYSA-N
Synonym triethyl orthoacetate, ethyl orthoacetate, ethane, 1,1,1-triethoxy, orthoacetic acid, triethyl ester, orthoacetic acid triethyl ester, triethoxyethane, triethoxy ethane, triethy orthoacetate, triethyl ortho-aceate, 1,1-triethoxyethane
PubChem CID 66221
IUPAC Name 1,1,1-triethoxyethane
SMILES CCOC(C)(OCC)OCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL06004AE
View Documents
Thermo Scientific Chemicals
L06004AE
100 mL
Each for $83.14
Only null left
 
AAL06004AP
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
L06004AP
500 mL
Each for $241.85
Only null left
 
AAL060040F
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
L060040F
2500 mL
Each for $727.30
Only null left
 
Description

Description

Triethyl orthoacetate is used as pharmaceutical and water scavenger. It is also used in organic synthesis to introduce the acetate group into an alcohol. It is involved in the Johnson-Claisen rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl orthoacetate is used as pharmaceutical and water scavenger. It is also used in organic synthesis to introduce the acetate group into an alcohol. It is involved in the Johnson-Claisen rearrangement.

Solubility
Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

78-39-7
162.229
NDQXKKFRNOPRDW-UHFFFAOYSA-N
66221
CCOC(C)(OCC)OCC
C8H18O3
MFCD00009223
triethyl orthoacetate, ethyl orthoacetate, ethane, 1,1,1-triethoxy, orthoacetic acid, triethyl ester, orthoacetic acid triethyl ester, triethoxyethane, triethoxy ethane, triethy orthoacetate, triethyl ortho-aceate, 1,1-triethoxyethane
1,1,1-triethoxyethane

Specifications

78-39-7
142°C to 143°C
C8H18O3
MFCD00009223
UN1993
Moisture sensitive
Slightly miscible with water.
CCOC(C)(OCC)OCC
162.229
162.23
Triethyl orthoacetate
0.887
32°C (89°F)
1.397
100 mL
506201
triethyl orthoacetate, ethyl orthoacetate, ethane, 1,1,1-triethoxy, orthoacetic acid, triethyl ester, orthoacetic acid triethyl ester, triethoxyethane, triethoxy ethane, triethy orthoacetate, triethyl ortho-aceate, 1,1-triethoxyethane
NDQXKKFRNOPRDW-UHFFFAOYSA-N
1,1,1-triethoxyethane
66221
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c

H226-H315-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 201-112-4

RTECSNumber : KJ4075000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only