missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethyl orthoformate, 98%, anhydrous, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 429278000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Triethyl orthoformate | |
0.8900g/mL | |
35°C | |
AcroSeal™ Glass Bottle | |
1.3900 to 1.3920 | |
800mL | |
15,698 | |
Solubility in water: 1.35g/L (20°C). Other solubilities: decomposes in water,soluble in alcohols,acetone,ethers,chloroform, | |
CCOC(OCC)OCC | |
148.2 | |
148.2 |
122-51-0 | |
146.0°C | |
97.5% min. (GC) | |
C7H16O3 | |
CH(OC2H5)3 | |
0.89 | |
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty | |
GKASDNZWUGIAMG-UHFFFAOYSA-N | |
diethoxymethoxyethane | |
31214 | |
98% |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 204-550-4
RUO â Research Use Only