missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethyl phosphate, 98+%, Thermo Scientific Chemicals

$98.25 - $298.58

Chemical Identifiers

CAS 78-40-0
Molecular Formula C6H15O4P
Molecular Weight (g/mol) 182.16
MDL Number MFCD00009077
InChI Key DQWPFSLDHJDLRL-UHFFFAOYSA-N
Synonym triethylphosphate, phosphoric acid, triethyl ester, tris ethyl phosphate, triethoxyphosphine oxide, triethylfosfat, ethyl phosphate eto 3po, triethylfosfat czech, ethyl phosphate van, unii-qih4k96k7j, phosphoric acid triethyl ester
PubChem CID 6535
ChEBI CHEBI:45927
IUPAC Name triethyl phosphate
SMILES CCOP(=O)(OCC)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1793836
View Documents
Thermo Scientific Chemicals
A1793836
500 g
Each for $98.25
 
AAA179380E
View Documents
Thermo Scientific Chemicals
A179380E
2500 g
Each for $298.58
 
Description

Description

Triethyl phosphate finds it major applications in plastics industry as a flame retardant, plasticizer and carrier, where it is available in the matrix. A further 10 to 20 % are used in other industrial branches as a solvent, plasticizer, flame retardant or intermediate for the production of pharmaceuticals, and lacquers. Triethyl phosphate is a useful synthetic intermediate used in the synthesis of mesoporous spheres of metal oxides and phosphates. It is also used as an industrial catalyst employed in ketene synthesis where the compound is hydrolyzed, as a polymer resin modifier, as a solvent, a car paint repairing product and as flame retarder

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl phosphate finds it major applications in plastics industry as a flame retardant, plasticizer and carrier, where it is available in the matrix. A further 10 to 20 % are used in other industrial branches as a solvent, plasticizer, flame retardant or intermediate for the production of pharmaceuticals, and lacquers. Triethyl phosphate is a useful synthetic intermediate used in the synthesis of mesoporous spheres of metal oxides and phosphates. It is also used as an industrial catalyst employed in ketene synthesis where the compound is hydrolyzed, as a polymer resin modifier, as a solvent, a car paint repairing product and as flame retarder

Solubility
Fully miscible in water at 20°C.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

78-40-0
182.16
DQWPFSLDHJDLRL-UHFFFAOYSA-N
6535
triethyl phosphate
C6H15O4P
MFCD00009077
triethylphosphate, phosphoric acid, triethyl ester, tris ethyl phosphate, triethoxyphosphine oxide, triethylfosfat, ethyl phosphate eto 3po, triethylfosfat czech, ethyl phosphate van, unii-qih4k96k7j, phosphoric acid triethyl ester
CHEBI:45927
CCOP(=O)(OCC)OCC

Specifications

78-40-0
215°C
Mild
1.405
500 g
triethylphosphate, phosphoric acid, triethyl ester, tris ethyl phosphate, triethoxyphosphine oxide, triethylfosfat, ethyl phosphate eto 3po, triethylfosfat czech, ethyl phosphate van, unii-qih4k96k7j, phosphoric acid triethyl ester
DQWPFSLDHJDLRL-UHFFFAOYSA-N
triethyl phosphate
6535
182.16
Triethyl phosphate
1.072
115°C (240°F)
C6H15O4P
MFCD00009077
14,9674
Fully miscible in water at 20°C.
CCOP(=O)(OCC)OCC
182.16
CHEBI:45927
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 201-114-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only