missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, 99%, pure

CAS: 121-44-8 | C6H15N | 101.19 g/mol

$55.38 - $439.53

Chemical Identifiers

CAS 121-44-8
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC157910050
SDS View Documents
Thermo Scientific Chemicals
157910050
5 mL Glass bottle
Each for $55.38
Only null left
 
AC157910010
SDS View Documents
Thermo Scientific Chemicals
157910010
1 L EcoSafPak™ Glass Bottle
Each for $230.41
Case of 4 Each for $0.00
Only null left
 
AC157910025
SDS View Documents
Thermo Scientific Chemicals
157910025
2.5 L Glass bottle
Each for $439.53
Case of 4 Each for $0.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-44-8
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
CHEBI:35026
CCN(CC)CC
ZMANZCXQSJIPKH-UHFFFAOYSA-N
8471
N,N-diethylethanamine

Specifications

121-44-8
0.7280g/mL
−11°C
98.5% min. (GC)
C6H15N
(C2H5)3N
04, 99
0.728
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
ZMANZCXQSJIPKH-UHFFFAOYSA-N
N,N-diethylethanamine
8471
CHEBI:35026
69mbar at 20°C
Pure
Triethylamine
-115.0°C
90.0°C
Authentic
Glass bottle
1.3990 to 1.4010
5 mL
01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336
15, 9831
Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride
CCN(CC)CC
101.19
0.36 mPa.s (20°C)
101.19
99%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS P Statement Highly flammable liquid and vapour. Toxic if inhaled. Toxic in contact with skin. Harmful if swallowed. Causes severe skin burns and eye damage. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Gently wash with plenty of soap and water. IF IN EYES: Rinse

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only