Learn More
Triethylamine hydrobromide, 98%, Thermo Scientific Chemicals
$78.55 - $263.01
Chemical Identifiers
CAS | 636-70-4 |
---|---|
Molecular Formula | C6H16BrN |
Molecular Weight (g/mol) | 182.11 |
MDL Number | MFCD00054287 |
InChI Key | NRTLTGGGUQIRRT-UHFFFAOYSA-N |
Synonym | triethylamine hydrobromide, ethanamine, n,n-diethyl-, hydrobromide, ethanamine, n,n-diethyl-, hydrobromide 1:1, n,n-diethylethanamine hydrobromide, acmc-20ajeg, et3n.hbr, triethylammonium bromide salt, triethylamine hydrobromide salt |
PubChem CID | 2734074 |
IUPAC Name | N,N-diethylethanamine;hydrobromide |
SMILES | [Br-].CC[NH+](CC)CC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1792622
|
Thermo Scientific Chemicals
A1792622 |
100 g |
Each for $78.55
|
|
|||||
AAA1792636
|
Thermo Scientific Chemicals
A1792636 |
500 g |
Each for $263.01
|
|
|||||
Description
Triethylamine hydrobromide is used to prepare dimethyl -alfa-diethylaminomaleate by reacting with dimethyl acetylenedicarbosylate. It is involved in the preparation of triphenylpropargylphosphonium bromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriethylamine hydrobromide is used to prepare dimethyl -alfa-diethylaminomaleate by reacting with dimethyl acetylenedicarbosylate. It is involved in the preparation of triphenylpropargylphosphonium bromide.
Solubility
Soluble in water, alcohol and chloroform.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
Chemical Identifiers
636-70-4 | |
182.11 | |
NRTLTGGGUQIRRT-UHFFFAOYSA-N | |
2734074 | |
[Br-].CC[NH+](CC)CC |
C6H16BrN | |
MFCD00054287 | |
triethylamine hydrobromide, ethanamine, n,n-diethyl-, hydrobromide, ethanamine, n,n-diethyl-, hydrobromide 1:1, n,n-diethylethanamine hydrobromide, acmc-20ajeg, et3n.hbr, triethylammonium bromide salt, triethylamine hydrobromide salt | |
N,N-diethylethanamine;hydrobromide |
Specifications
636-70-4 | |
Amine-like | |
(CH3CH2)3N·HBr | |
100 g | |
triethylamine hydrobromide, ethanamine, n,n-diethyl-, hydrobromide, ethanamine, n,n-diethyl-, hydrobromide 1:1, n,n-diethylethanamine hydrobromide, acmc-20ajeg, et3n.hbr, triethylammonium bromide salt, triethylamine hydrobromide salt | |
NRTLTGGGUQIRRT-UHFFFAOYSA-N | |
N,N-diethylethanamine;hydrobromide | |
2734074 | |
98% |
∼250°C (decomposition) | |
C6H16BrN | |
MFCD00054287 | |
Hygroscopic | |
Soluble in water,alcohol and chloroform. | |
[Br-].CC[NH+](CC)CC | |
182.11 | |
182.1 | |
Triethylamine hydrobromide |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 211-263-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only