missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethylamine Phosphate 96.0+%, TCI America™
Supplier: TCI America T130025G
Specifications
Triethylamine Phosphate | |
White | |
MFCD00067475 | |
triethylamine phosphate, triethylammonium phosphate, triethylamine phosphate 1:1, n,n-diethylethanamine; phosphoric acid, phosphoric acid; triethylamine, ethanamine, n,n-diethyl-, phosphate, ethanamine, n,n-diethyl-, phosphate 1:?, ethanamine, n,n-diethyl-, phosphate 1:1, teap, acmc-20aj7n | |
OP(O)(O)=O.CCN(CC)CC | |
199.19 | |
199.19 | |
Crystalline Powder |
35365-94-7 | |
C6H18NO4P | |
25 g | |
UNXNGGMLCSMSLH-UHFFFAOYSA-N | |
phosphoric acid; triethylamine | |
61946 | |
≥96.0% (T) |
Chemical Identifiers
35365-94-7 | |
199.19 | |
UNXNGGMLCSMSLH-UHFFFAOYSA-N | |
61946 | |
OP(O)(O)=O.CCN(CC)CC |
C6H18NO4P | |
MFCD00067475 | |
triethylamine phosphate, triethylammonium phosphate, triethylamine phosphate 1:1, n,n-diethylethanamine; phosphoric acid, phosphoric acid; triethylamine, ethanamine, n,n-diethyl-, phosphate, ethanamine, n,n-diethyl-, phosphate 1:?, ethanamine, n,n-diethyl-, phosphate 1:1, teap, acmc-20aj7n | |
phosphoric acid; triethylamine |
Safety and Handling
RTECSNumber : YE0175000
TSCA : Yes