missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylborane, 1M soln. in THF, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L15109AE

 View more versions of this product

Catalog No. AAL15109AE


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylborane is catalyst for allylation of aldehydes, decarboxylative C-C bond cleavage reactions, rhenium hydride / boron Lewis acid cocatalysis of alkene hydrogenations, Regioselective hydroxyalkylation of unsaturated oxime ethers. Reactant for radical reductions of alkyl bromides with N-heterocyclic carbene boranes and synthesis of tetramethylammonium trialkylphenylborate salts with oxidation potential.

Solubility
Reacts with water.

Notes
Air Sensitive, store away form air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Triethylborane
0.865
C6H15B
MFCD00009022
UN2924
Air Sensitive
Reacts with water.
CCB(CC)CC
98.00
98
97-94-9
−17°C (1°F)
(CH3CH2)3B
100 mL
1731462
triethylboron, borane, triethyl, triethylborine, boron triethyl, boron ethyl, unii-z3s980z4p3, hsdb 897, triethylboran, borethyl, triethyl borane
LALRXNPLTWZJIJ-UHFFFAOYSA-N
triethylborane
7357
1M soln. in THF

Chemical Identifiers

97-94-9
98.00
LALRXNPLTWZJIJ-UHFFFAOYSA-N
7357
CCB(CC)CC
C6H15B
MFCD00009022
triethylboron, borane, triethyl, triethylborine, boron triethyl, boron ethyl, unii-z3s980z4p3, hsdb 897, triethylboran, borethyl, triethyl borane
triethylborane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H332-H314-H335-H336-H351

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

EINECSNumber : 202-620-9

RTECSNumber : ED2100000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only