missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethylchlorogermane 97.0+%, TCI America™
Supplier: TCI America T11601G
Specifications
Triethylchlorogermane | |
Yellow | |
C6H17ClGe | |
1 g | |
FJAWDWVPBHGANJ-UHFFFAOYSA-N | |
chloro(triethyl)germane | |
70438 | |
≥97.0% (GC) |
994-28-5 | |
176°C | |
MFCD00013591 | |
triethylgermanium chloride, triethylchlorogermane, chloro triethyl germane, triethylchlorogermene, ccris 2650, triethylgermyl chloride, germanium triethyl chloride, acmc-209sco, chlorotriethylgermane | |
Cl.CC[GeH](CC)CC | |
197.28 | |
195.27 | |
Liquid |
Chemical Identifiers
994-28-5 | |
197.28 | |
FJAWDWVPBHGANJ-UHFFFAOYSA-N | |
70438 | |
Cl.CC[GeH](CC)CC |
C6H17ClGe | |
MFCD00013591 | |
triethylgermanium chloride, triethylchlorogermane, chloro triethyl germane, triethylchlorogermene, ccris 2650, triethylgermyl chloride, germanium triethyl chloride, acmc-209sco, chlorotriethylgermane | |
chloro(triethyl)germane |
Safety and Handling
TSCA : No