missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylene glycol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A10772AP

 View more versions of this product

Catalog No. AAA10772AP

Explore available promotions

Description

Description

Triethylene glycol is used as a plasticizer, as an additive for hydraulic fluids and brake fluids, and as a disinfectant. It is an active component of certain pigments, printing dyes, inks and paste. It finds application as a liquid desiccant and used in the dehydration of natural gas, carbon dioxide, hydrogen sulfide and air conditioning systems. It plays as an important role in anti-freeze and de-icing products, cleaning and furnishing care products, lubricant and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylene glycol is used as a plasticizer, as an additive for hydraulic fluids and brake fluids, and as a disinfectant. It is an active component of certain pigments, printing dyes, inks and paste. It finds application as a liquid desiccant and used in the dehydration of natural gas, carbon dioxide, hydrogen sulfide and air conditioning systems. It plays as an important role in anti-freeze and de-icing products, cleaning and furnishing care products, lubricant and greases.

Solubility
Miscible with water, ethanol, acetone, benzene, toluene, acetic acid, glycerine, pyridine and aldehydes. Slightly miscible with diethyl ether and chloroform. Immiscible with oil, fat, petroleum ether and most hydrocarbons.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acids and bases.
Specifications

Specifications

Triethylene glycol
°C to 7°C
284°C to 288°C
C6H14O4
H(OCH2CH2)3OH
500 mL
Hygroscopic
triethylene glycol, triglycol, 2,2'-ethane-1,2-diylbis oxy diethanol, trigen, triethylenglykol, triethyleneglycol, 2,2'-ethylenedioxydiethanol, 1,2-bis 2-hydroxyethoxy ethane, 3,6-dioxaoctane-1,8-diol, 2,2'-ethylenedioxy diethanol
ZIBGPFATKBEMQZ-UHFFFAOYSA-N
2-[2-(2-hydroxyethoxy)ethoxy]ethanol
8172
150.17
Liquid
112-27-6
1.125 g/mL
165°C (329°F)
1.4555
MFCD00002880,MFCD00081839,MFCD01779596,MFCD01779599,MFCD01779601,MFCD01779603,MFCD01779605,MFCD01779609,MFCD01779611,MFCD01779612,MFCD01779614,MFCD01779615,MFCD01779616
969357
14,9670
Miscible with water,ethanol,acetone,benzene,toluene,acetic acid,glycerine,pyridine and aldehydes. Slightly miscible with diethyl ether and chloroform. Immiscible with oil,fat,petroleum ether and most hydrocarbons.
OCCOCCOCCO
150.17
CHEBI:44926
99%

Chemical Identifiers

112-27-6
150.17
ZIBGPFATKBEMQZ-UHFFFAOYSA-N
8172
2-[2-(2-hydroxyethoxy)ethoxy]ethanol
C6H14O4
MFCD00002880,MFCD00081839,MFCD01779596,MFCD01779599,MFCD01779601,MFCD01779603,MFCD01779605,MFCD01779609,MFCD01779611,MFCD01779612,MFCD01779614,MFCD01779615,MFCD01779616
triethylene glycol, triglycol, 2,2'-ethane-1,2-diylbis oxy diethanol, trigen, triethylenglykol, triethyleneglycol, 2,2'-ethylenedioxydiethanol, 1,2-bis 2-hydroxyethoxy ethane, 3,6-dioxaoctane-1,8-diol, 2,2'-ethylenedioxy diethanol
CHEBI:44926
OCCOCCOCCO
Safety and Handling

Safety and Handling

EINECSNumber : 203-953-2

RTECSNumber : YE4550000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only