missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylenetetramine-N,N,N',N″,N‴,N‴-hexaacetic acid, 98%, Thermo Scientific Chemicals

$115.78 - $511.86

Chemical Identifiers

CAS 869-52-3
Molecular Formula C18H30N4O12
Molecular Weight (g/mol) 494.454
MDL Number MFCD00004290
InChI Key RAEOEMDZDMCHJA-UHFFFAOYSA-N
Synonym ttha, hexaacetic acid, triethylenetetraminehexaacetate, 3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl, triethylenetetranitrilohexaacetic acid, triethylenetetramine-n,n,n',n,n',n'-hexaacetic acid, triethylenetetramine-n,n,n',n',n,n-hexaacetic acid, triethylenetetramine-n,n,n',n,n', 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecane-1,14-dioic acid
PubChem CID 70088
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-[2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]ethyl]amino]acetic acid
SMILES C(CN(CCN(CC(=O)O)CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4473704
View Documents
Thermo Scientific Chemicals
04473704
2 g
Each for $115.78
 
AA4473709
View Documents
Thermo Scientific Chemicals
04473709
10 g
Each for $511.86
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

869-52-3
494.454
RAEOEMDZDMCHJA-UHFFFAOYSA-N
70088
C(CN(CCN(CC(=O)O)CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O
C18H30N4O12
MFCD00004290
ttha, hexaacetic acid, triethylenetetraminehexaacetate, 3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl, triethylenetetranitrilohexaacetic acid, triethylenetetramine-n,n,n',n,n',n'-hexaacetic acid, triethylenetetramine-n,n,n',n',n,n-hexaacetic acid, triethylenetetramine-n,n,n',n,n', 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecane-1,14-dioic acid
2-[2-[bis(carboxymethyl)amino]ethyl-[2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]ethyl]amino]acetic acid

Specifications

869-52-3
MFCD00004290
ttha, hexaacetic acid, triethylenetetraminehexaacetate, 3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl, triethylenetetranitrilohexaacetic acid, triethylenetetramine-n,n,n',n,n',n'-hexaacetic acid, triethylenetetramine-n,n,n',n',n,n-hexaacetic acid, triethylenetetramine-n,n,n',n,n', 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecanedioic acid, 3,6,9,12-tetrakis carboxymethyl-3,6,9,12-tetraazatetradecane-1,14-dioic acid
C(CN(CCN(CC(=O)O)CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O
494.454
494.45
Powder
C18H30N4O12
2 g
RAEOEMDZDMCHJA-UHFFFAOYSA-N
2-[2-[bis(carboxymethyl)amino]ethyl-[2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]ethyl]amino]acetic acid
70088
98%
Triethylenetetramine-N,N,N',N'',N''',N'''-hexaacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 212-790-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only