missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethylenetetramine Tetrahydrochloride 60.0+%, TCI America™
Supplier: TCI America T121225G
Specifications
Triethylenetetramine Tetrahydrochloride | |
C6H22Cl4N4 | |
25 g | |
OKHMDSCYUWAQPT-UHFFFAOYSA-N | |
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine;tetrahydrochloride | |
71434 | |
≥60.0% (GC) |
4961-40-4 | |
MFCD00012890 | |
triethylenetetramine tetrahydrochloride, trientine tetrahydrochloride, unii-7360ure56q, trien tetrahydrochloride, 1,2-ethanediamine, n,n'-bis 2-aminoethyl-, tetrahydrochloride, triethylenetetramine, tetrahydrochloride, 3,6-diazaoctane-1,8-diamine tetrahydrochloride, n,n'-bis 2-aminoethyl-1,2-ethanediamine tetrahydrochloride, 1,2-ethanediamine, n1,n2-bis 2-aminoethyl-, hydrochloride 1:4, n1,n1'-ethane-1,2-diyl bis ethane-1,2-diamine tetrahydrochloride | |
C(CNCCNCCN)N.Cl.Cl.Cl.Cl | |
292.07 | |
292.07 | |
Crystalline Powder |
Chemical Identifiers
4961-40-4 | |
292.07 | |
OKHMDSCYUWAQPT-UHFFFAOYSA-N | |
71434 | |
C(CNCCNCCN)N.Cl.Cl.Cl.Cl |
C6H22Cl4N4 | |
MFCD00012890 | |
triethylenetetramine tetrahydrochloride, trientine tetrahydrochloride, unii-7360ure56q, trien tetrahydrochloride, 1,2-ethanediamine, n,n'-bis 2-aminoethyl-, tetrahydrochloride, triethylenetetramine, tetrahydrochloride, 3,6-diazaoctane-1,8-diamine tetrahydrochloride, n,n'-bis 2-aminoethyl-1,2-ethanediamine tetrahydrochloride, 1,2-ethanediamine, n1,n2-bis 2-aminoethyl-, hydrochloride 1:4, n1,n1'-ethane-1,2-diyl bis ethane-1,2-diamine tetrahydrochloride | |
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine;tetrahydrochloride |
Safety and Handling
RTECSNumber : KH8587087
TSCA : Yes