missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylsilyl trifluoromethanesulfonate, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 312700100

 View more versions of this product

Catalog No. AC312700100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Silylation reagent
Specifications

Specifications

Triethylsilyl trifluoromethanesulfonate
1.1600g/mL
72°C
99%
C7H15F3O3SSi
CF3SO3Si(C2H5)3
10g
triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf
STMPXDBGVJZCEX-UHFFFAOYSA-N
triethylsilyl trifluoromethanesulfonate
2733308
99%
79271-56-0
85.0°C to 86.0°C (12.0mmHg)
Authentic
Glass bottle
1.3890 to 1.3920
MFCD00000407
1.16
Solubility in water: soluble
CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F
264.33
264.34

Chemical Identifiers

79271-56-0
264.33
STMPXDBGVJZCEX-UHFFFAOYSA-N
2733308
CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F
C7H15F3O3SSi
MFCD00000407
triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf
triethylsilyl trifluoromethanesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 279-124-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only