missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilyl Trifluoromethanesulfonate 98.0+%, TCI America™

$80.16 - $227.41

Chemical Identifiers

CAS 79271-56-0
Molecular Formula C7H15F3O3SSi
Molecular Weight (g/mol) 264.33
MDL Number MFCD00000407
InChI Key STMPXDBGVJZCEX-UHFFFAOYSA-N
Synonym triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf
PubChem CID 2733308
IUPAC Name triethylsilyl trifluoromethanesulfonate
SMILES CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T16895G
View Documents
TCI America
T16895G
5 g
Each for $80.16
Only null left
 
T168925G
View Documents
TCI America
T168925G
25 g
Each for $227.41
Only null left
 
Description

Description

Specifications

Chemical Identifiers

79271-56-0
264.33
STMPXDBGVJZCEX-UHFFFAOYSA-N
2733308
CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F
C7H15F3O3SSi
MFCD00000407
triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf
triethylsilyl trifluoromethanesulfonate

Specifications

79271-56-0
79°C
MFCD00000407
1760
STMPXDBGVJZCEX-UHFFFAOYSA-N
triethylsilyl trifluoromethanesulfonate
2733308
≥98.0% (T)
Triethylsilyl Trifluoromethanesulfonate
Yellow
C7H15F3O3SSi
5 g
triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf
CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F
264.33
264.33
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'