Learn More
Triethylsulfonium iodide, 97%, Thermo Scientific Chemicals
$86.74 - $1082.92
Chemical Identifiers
CAS | 1829-92-1 |
---|---|
Molecular Formula | C6H15IS |
Molecular Weight (g/mol) | 246.15 |
MDL Number | MFCD00051867 |
InChI Key | LKDQWVKWYGOVJW-UHFFFAOYSA-M |
Synonym | triethylsulfonium iodide, triethylsulfanium iodide, triethylsulphonium iodide, sulfonium, triethyl-, iodide, acmc-20aje7, sulfonium,triethyl-,iodide |
PubChem CID | 74589 |
IUPAC Name | triethylsulfanium;iodide |
SMILES | CC[S+](CC)CC.[I-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0066706
|
Thermo Scientific Chemicals
L0066706 |
5 g |
Each for $86.74
|
|
|||||
AAL0066714
|
Thermo Scientific Chemicals
L0066714 |
25 g |
Each for $299.49
|
|
|||||
AAL0066722
|
Thermo Scientific Chemicals
L0066722 |
100 g |
Each for $1,082.92
|
|
|||||
Description
Triethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.
Solubility
It is soluble in water.
Notes
Light, Heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Chemical Identifiers
1829-92-1 | |
246.15 | |
LKDQWVKWYGOVJW-UHFFFAOYSA-M | |
74589 | |
CC[S+](CC)CC.[I-] |
C6H15IS | |
MFCD00051867 | |
triethylsulfonium iodide, triethylsulfanium iodide, triethylsulphonium iodide, sulfonium, triethyl-, iodide, acmc-20aje7, sulfonium,triethyl-,iodide | |
triethylsulfanium;iodide |
Specifications
1829-92-1 | |
C6H15IS | |
5 g | |
Light sensitive | |
It is soluble in water. | |
CC[S+](CC)CC.[I-] | |
246.15 | |
246.15 | |
Triethylsulfonium iodide |
∽148°C (decomposition) | |
MFCD00051867 | |
3911790 | |
triethylsulfonium iodide, triethylsulfanium iodide, triethylsulphonium iodide, sulfonium, triethyl-, iodide, acmc-20aje7, sulfonium,triethyl-,iodide | |
LKDQWVKWYGOVJW-UHFFFAOYSA-M | |
triethylsulfanium;iodide | |
74589 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 217-383-7
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only