Learn More
Trifluoroacetaldehyde methyl hemiacetal, tech. 90%, Thermo Scientific Chemicals
$72.52 - $72.52
Chemical Identifiers
CAS | 431-46-9 |
---|---|
Molecular Formula | C3H5F3O2 |
Molecular Weight (g/mol) | 130.07 |
MDL Number | MFCD00013572 |
InChI Key | GWTBCUWZAVMAQV-UHFFFAOYNA-N |
Synonym | trifluoroacetaldehyde methyl hemiacetal, fluoral methyl hemiacetal, 1-methoxy-2,2,2-trifluoroethanol, ethanol, 2,2,2-trifluoro-1-methoxy, trifluoroacetaldehyde methyl hemiacetal, tech., trifluoroacetaldehyde hemi methyl acetal, 2,2,2-erifluoro-1-methoxyethanol, ethanol, 1-methoxy-2,2,2-trifluoro, zlchem 1308, fluoral hemimethylacetal |
PubChem CID | 9892 |
SMILES | COC(O)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1836906
|
Thermo Scientific Chemicals
A1836906 |
5 g |
Each for $72.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
431-46-9 | |
130.07 | |
GWTBCUWZAVMAQV-UHFFFAOYNA-N | |
9892 |
C3H5F3O2 | |
MFCD00013572 | |
trifluoroacetaldehyde methyl hemiacetal, fluoral methyl hemiacetal, 1-methoxy-2,2,2-trifluoroethanol, ethanol, 2,2,2-trifluoro-1-methoxy, trifluoroacetaldehyde methyl hemiacetal, tech., trifluoroacetaldehyde hemi methyl acetal, 2,2,2-erifluoro-1-methoxyethanol, ethanol, 1-methoxy-2,2,2-trifluoro, zlchem 1308, fluoral hemimethylacetal | |
COC(O)C(F)(F)F |
Specifications
431-46-9 | |
96°C | |
C3H5F3O2 | |
MFCD00013572 | |
UN3271 | |
trifluoroacetaldehyde methyl hemiacetal, fluoral methyl hemiacetal, 1-methoxy-2,2,2-trifluoroethanol, ethanol, 2,2,2-trifluoro-1-methoxy, trifluoroacetaldehyde methyl hemiacetal, tech., trifluoroacetaldehyde hemi methyl acetal, 2,2,2-erifluoro-1-methoxyethanol, ethanol, 1-methoxy-2,2,2-trifluoro, zlchem 1308, fluoral hemimethylacetal | |
COC(O)C(F)(F)F | |
130.07 | |
130.07 | |
Technical |
1.36 | |
42°C (107°F) | |
1.333 | |
5 g | |
1737758 | |
GWTBCUWZAVMAQV-UHFFFAOYNA-N | |
2,2,2-trifluoro-1-methoxyethanol | |
9892 | |
90% | |
Trifluoroacetaldehyde methyl hemiacetal |
Safety and Handling
GHS H Statement
H226-H302
Flammable liquid and vapour.
Harmful if swallowed.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H226-H302
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber : 207-072-4
RTECSNumber : KL6600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only