Learn More
Trifluoroacetic acid, 99.5%, for biochemistry, Thermo Scientific Chemicals
$96.25 - $1,913.51
Chemical Identifiers
CAS | 76-05-1 |
---|---|
Molecular Formula | C2HF3O2 |
Molecular Weight (g/mol) | 114.02 |
MDL Number | MFCD00004169 |
InChI Key | DTQVDTLACAAQTR-UHFFFAOYSA-N |
Synonym | trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova |
PubChem CID | 6422 |
ChEBI | CHEBI:45892 |
IUPAC Name | trifluoroacetic acid |
SMILES | OC(=O)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC293810250
|
Thermo Scientific Chemicals
293810250 |
25 mL | Glass bottle |
N/A
|
|
||||
AC293811000
|
Thermo Scientific Chemicals
293811000 |
100 mL | Glass bottle |
N/A
|
|
||||
AC293812500
|
Thermo Scientific Chemicals
293812500 |
250 mL | Glass bottle |
N/A
|
|
||||
AC293810010
|
Thermo Scientific Chemicals
293810010 |
1 L | Glass bottle |
N/A
|
|
||||
AC293810025
|
Thermo Scientific Chemicals
293810025 |
2.5 L | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistryChemical Identifiers
76-05-1 | |
114.02 | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
6422 | |
trifluoroacetic acid |
C2HF3O2 | |
MFCD00004169 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
CHEBI:45892 | |
OC(=O)C(F)(F)F |
Specifications
-15.0°C | |
72.0°C | |
C2HF3O2 | |
MFCD00004169 | |
01,1219; 02,433; 03,305; 04,530; 05,695; 06,613; 07,388; 08,503; 09,483; 10,418; 11,557; 12,529; 14,322; 15,338; 16,83; 17,340 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
trifluoroacetic acid | |
6422 | |
114.02 | |
Biochemical | |
25 mL | |
Glass bottle | |
1.3 | |
Trifluoroacetic acid |
76-05-1 | |
99.5% | |
CF3CO2H | |
02, II, 186 | |
15, 9847 | |
Solubility in water: miscible. Other solubilities: miscible with ether, acetone, ethanol, benzene,, ccl4, hexane, methanol, xylene and hexane | |
OC(=O)C(F)(F)F | |
114.02 | |
CHEBI:45892 | |
99.5% | |
1.5350g/mL | |
Authentic | |
Liquid | |
1.535 |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only