missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trifluoroacetic Acid, 99.9%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor TR1191KG

 View more versions of this product

Catalog No. 18614559


Explore available promotions

Description

Description

Specifications

Specifications

76-05-1
99.9%
1 kg
1
Reagent
Amber Glass Bottle

Chemical Identifiers

76-05-1
Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.