missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trifluoroacetic Acid, 99.9%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor TR119500GM
Specifications
76-05-1 | |
99.9% | |
500 g |
1 | |
Reagent | |
Amber Glass Bottle |
Chemical Identifiers
76-05-1 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.