missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trifluoroacetic acid, Eluent additive for LC-MS, Honeywell Fluka™
Eluent additive for LC-MS
Supplier: Honeywell Chemicals 409675X10ML
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
max. 0.025 at 320 nm, max. 0.03 at 300 nm, max. 0.04 at 290 nm, max. 0.05 280 nm, max. 0.1 at 270 nm, max. 0.9 at 260 nm | |
Eluent additive for LC-MS | |
76-05-1 | |
100.0000% | |
CF3COOH | |
UN2699 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
OC(=O)C(F)(F)F | |
114.02 | |
CHEBI:45892 | |
>99% | |
1.489Â g/mLÂ at 20Â °C(lit.) | |
100°C | |
3.9 (vs air) |
Trifluoroacetic acid | |
-15.4°C (literature) | |
72°C | |
C2HF3O2 | |
MFCD00004169 | |
742035 | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
2,2,2-trifluoroacetic acid | |
6422 | |
114.02 | |
LC-MS | |
5 x 10 mL | |
Ampoule | |
97.5 mmHg (20°C) |
Chemical Identifiers
76-05-1 | |
114.02 | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
6422 | |
OC(=O)C(F)(F)F |
C2HF3O2 | |
MFCD00004169 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
CHEBI:45892 |
Safety and Handling
P273-P280-P305 + P351 + P338-P310
ShelfLife : 360 days from date of manufacture
DOTInformation : Transport Hazard Class: 8; Packing Group:3: I; Proper Shipping Name: Trifluoroacetic acid
EINECSNumber : 200-929-3
RTECSNumber : 200-929-3
Recommended Storage : Room Temp