missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trifluoroacetic Acid (For Peptide and Protein Analysis/Certified), Fisher Chemical

Supplier:  Fisher Chemical O4902100

Catalog No. O4902-100



Explore available promotions

Description

Description

Specifications

Specifications

Trifluoroacetic Acid
For Peptide and Protein Analysis
76-05-1
Colorless
C2HF3O2
MFCD00004169
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
DTQVDTLACAAQTR-UHFFFAOYSA-N
trifluoroacetic acid
6422
114.02
Certified
100 mL
Liquid
0.45
-15°C
72°C
≥98.5 %
CF3COOH
2699
Miscible in water
OC(=O)C(F)(F)F
114.02
CHEBI:45892
≥98.5%
1.535g/cm³
Glass Bottle
107mbar at 25°C

Chemical Identifiers

76-05-1
114.02
DTQVDTLACAAQTR-UHFFFAOYSA-N
6422
trifluoroacetic acid
C2HF3O2
MFCD00004169
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
CHEBI:45892
OC(=O)C(F)(F)F
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Causes severe burns by all exposure routes. Inhalation may cause central nervous system effects. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:0
Instability:0

DOTInformation : Class 8: Corrosive

SDS Documents

Documents

Promotions

Promotions