Learn More
Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™
Trifluoroacetic Acid, >-99.5%, C2HF3O2, CAS Number-76-05-1, trifluoroethanoic acid, trifluoroacetic acid, cf3cooh, acetic acid, trifluoro, trifluoro-acetic acid, perfluoroacetic acid, kyselina trifluoroctova, trifluoro acetic acid, trifluoroacetic acid, trifluoroaceticacid, 50mL
Supplier: Fisher Chemical A11605AMP
Description
Trifluoroacetic Acid, >-99.5%, C2HF3O2, CAS Number-76-05-1, trifluoroethanoic acid, trifluoroacetic acid, cf3cooh, acetic acid, trifluoro, trifluoro-acetic acid, perfluoroacetic acid, kyselina trifluoroctova, trifluoro acetic acid, trifluoroacetic acid, trifluoroaceticacid, 50mL
- Ideal high purity mobile phase additive for LC/MS and UHPLC-UV applications
- Evaluated for metal impurities at ppb concentrations for minimal metal mass adduct formation
- Screened for UV-absorbing contaminants to reduce interferences and for smoother baselines
- TFA is a common mobile phase additive that can function as an ion pair reagent
Specifications
0.003 AU max. at 254nm, 0.03 AU max. at 300nm, 0.06 AU max. at 230nm, 0.06 AU max. at 275nm, 0.18 AU max. at 220nm, 0.55 AU max. at 210nm | |
0.45 | |
76-05-1 | |
Colorless | |
C2HF3O2 | |
MFCD00004169 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
OC(=O)C(F)(F)F | |
114.02 | |
CHEBI:45892 | |
≥99.5% | |
1.535g/cm³ | |
Ampule | |
1ppm max. | |
107mbar 25 |
Trifluoroacetic Acid | |
-15°C | |
72°C | |
≥99.5 % | |
CF3COOH | |
15, 9847 | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
trifluoroacetic acid | |
6422 | |
114.02 | |
OPTIMA™ LC-MS | |
0.5 mL | |
Liquid | |
3.9 |
Chemical Identifiers
76-05-1 | |
114.02 | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
6422 | |
trifluoroacetic acid |
C2HF3O2 | |
MFCD00004169 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
CHEBI:45892 | |
OC(=O)C(F)(F)F |
Safety and Handling
DANGER!
Emergency Overview
Causes severe burns by all exposure routes. Inhalation may cause central nervous system effects. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.
NFPA
Health:3
Flammability:0
Instability:0
DOTInformation : DOT Class 8, : Corrosive