missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals

Trifluoroacetic anhydride, 99+%, Acros Organics, Quantity: 500g, Packaging: Glass Bottle, Boiling Point: 39.1 deg.C, Undesignated, Melting Point: -63.5 deg.C, Molecular Weight: 210.03, Percent Purity: 99+%, Assay Percent Range: 99% min. (GC), Beilstein: 02, II, 186, CAS: 407-25-0

$60.32 - $346.90

Chemical Identifiers

CAS 407-25-0
Molecular Formula C4F6O3
Molecular Weight (g/mol) 210.03
MDL Number MFCD00000416
InChI Key QAEDZJGFFMLHHQ-UHFFFAOYSA-N
Synonym trifluoroacetic anhydride, trifluoroacetic acid anhydride, 2,2,2-trifluoroacetic anhydride, bis trifluoroacetic anhydride, acetic acid, trifluoro-, anhydride, perfluoroacetic anhydride, trifluoroacetyl anhydride, tfaa, hexafluoroacetic anhydride, anhydrid kyseliny trifluoroctove
PubChem CID 9845
IUPAC Name (2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate
SMILES FC(F)(F)C(=O)OC(=O)C(F)(F)F
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC147810250
SDS View Documents
Thermo Scientific Chemicals
147810250
25 g Glass Bottle
Each for $60.32
Case of 6 Each for $361.92
 
AC147811000
SDS View Documents
Thermo Scientific Chemicals
147811000
100 g Glass Bottle
Each for $109.80
Case of 6 Each for $658.80
 
AC147812501
SDS View Documents
Thermo Scientific Chemicals
147812501
250 g Glass bottle
Each for $230.14
 
AC147815000
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
147815000
500 g Glass Bottle
Each for $346.90
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
Specifications

Chemical Identifiers

407-25-0
210.03
QAEDZJGFFMLHHQ-UHFFFAOYSA-N
9845
FC(F)(F)C(=O)OC(=O)C(F)(F)F
C4F6O3
MFCD00000416
trifluoroacetic anhydride, trifluoroacetic acid anhydride, 2,2,2-trifluoroacetic anhydride, bis trifluoroacetic anhydride, acetic acid, trifluoro-, anhydride, perfluoroacetic anhydride, trifluoroacetyl anhydride, tfaa, hexafluoroacetic anhydride, anhydrid kyseliny trifluoroctove
(2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate

Specifications

407-25-0
1.5100g/mL
Authentic
Glass Bottle
1.3
MFCD00000416
02, II, 186
trifluoroacetic anhydride, trifluoroacetic acid anhydride, 2,2,2-trifluoroacetic anhydride, bis trifluoroacetic anhydride, acetic acid, trifluoro-, anhydride, perfluoroacetic anhydride, trifluoroacetyl anhydride, tfaa, hexafluoroacetic anhydride, anhydrid kyseliny trifluoroctove
QAEDZJGFFMLHHQ-UHFFFAOYSA-N
(2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate
9845
141mbar at 20°C
Liquid
-63.5°C
39.1°C
99+%
C4F6O3
(CF3CO)2O
25 g
01,1221; 03,308; 05,701; 06,616; 07,389; 09,484; 12,530; 14,151; 15,14; 01,373
Solubility in water: hydrolysis. Other solubilities: miscible with polar organic solvents, partically soluble in unpolar hydrocarbons
FC(F)(F)C(=O)OC(=O)C(F)(F)F
210.03
210.03
≥99%
Trifluoroacetic anhydride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minu

GHS Signal Word: Danger

EINECSNumber : 206-982-9

SDS
Documents

Documents

RUO – Research Use Only