Learn More
Trifluoromethanesulfonic anhydride, 98%, Thermo Scientific Chemicals
Alfa Aesar Trifluoromethanesulfonic anhydride, 98%, Quantity: 250g, Boiling Point: 81 deg.C to 83 deg.C, Melting Point: -80 deg.C, Molecular Weight: 282.127, Percent Purity: 98%, Beilstein: 1813600, CAS: 358-23-6, ChEBI: CHEBI:48509, Density: 1.72
Supplier: Thermo Scientific Chemicals A1176718
Description
Trifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTrifluoromethanesulfonic anhydride is used to convert phenols and imine into triflic ester and NTf group. It is a strong electrophile used for the introduction of triflyl group in chemical synthesis. It serves as a reagent in the preparation of alkyl and vinyl triflates, and for the stereoselective synthesis of mannosazide methyl uronate donors. It acts as a catalyst for glycosylation with anomeric hydroxy sugars to prepare polysaccharides.
Solubility
Miscible with dichloromethane. Immiscible with hydrocarbons.
Notes
Moisture sensitive. Incompatible with water, strong bases, strong oxidizing agents, acids and alcohols.
Specifications
Trifluoromethanesulfonic anhydride | |
-80°C | |
81°C to 83°C | |
1.321 | |
MFCD00000408 | |
UN3265 | |
Moisture sensitive | |
Miscible with dichloromethane. Immiscible with hydrocarbons. | |
C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F | |
282.127 | |
CHEBI:48509 | |
98% |
358-23-6 | |
1.72 | |
C2F6O5S2 | |
(CF3SO2)2O | |
50 g | |
1813600 | |
trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride | |
WJKHJLXJJJATHN-UHFFFAOYSA-N | |
trifluoromethylsulfonyl trifluoromethanesulfonate | |
67749 | |
282.13 |
Chemical Identifiers
358-23-6 | |
282.127 | |
WJKHJLXJJJATHN-UHFFFAOYSA-N | |
67749 | |
trifluoromethylsulfonyl trifluoromethanesulfonate |
C2F6O5S2 | |
MFCD00000408 | |
trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride | |
CHEBI:48509 | |
C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: I
EINECSNumber : 206-616-8
RTECSNumber : PB2772000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only