missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trifluoromethanesulfonic Anhydride 98.0+%, TCI America™
$93.43 - $783.73
Chemical Identifiers
CAS | 358-23-6 |
---|---|
Molecular Formula | C2F6O5S2 |
Molecular Weight (g/mol) | 282.127 |
MDL Number | MFCD00000408 |
InChI Key | WJKHJLXJJJATHN-UHFFFAOYSA-N |
Synonym | trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride |
PubChem CID | 67749 |
ChEBI | CHEBI:48509 |
IUPAC Name | trifluoromethylsulfonyl trifluoromethanesulfonate |
SMILES | C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F |
Chemical Identifiers
358-23-6 | |
282.127 | |
WJKHJLXJJJATHN-UHFFFAOYSA-N | |
67749 | |
trifluoromethylsulfonyl trifluoromethanesulfonate |
C2F6O5S2 | |
MFCD00000408 | |
trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride | |
CHEBI:48509 | |
C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F |
Specifications
358-23-6 | |
Colorless | |
C2F6O5S2 | |
10 g | |
trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride | |
C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F | |
282.127 | |
CHEBI:48509 | |
≥98.0% (T) | |
Trifluoromethanesulfonic Anhydride |
-80°C | |
84°C | |
MFCD00000408 | |
3265 | |
WJKHJLXJJJATHN-UHFFFAOYSA-N | |
trifluoromethylsulfonyl trifluoromethanesulfonate | |
67749 | |
282.13 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-3807
TSCA : Yes
Recommended Storage : Refrigerator